Learn More
Medchemexpress LLC Cyclo(Arg-Gly-Asp-D-Tyr-Lys) trifluoroacetate | 250612-42-1 | MFCD08459533 | 99.9% | 847.72 g/mol | C31H43F6N9O12 | 25 MG
SDP
Catalog No. 5000212790
Encompass_Preferred
Cyclo(RGDyK) trifluoroacetate is a cyclic pentapeptide integrin inhibitor supplied as the trifluoroacetate salt. It selectively binds the αVβ3 integrin (IC50 ≈ 20 nM) and is provided as a high-purity white to off-white solid for biochemical and cell-based research. For research use only; not for human or clinical use.
  • Selective αVβ3 integrin inhibition with low-nanomolar potency.
  • High chemical purity (~99.9%) suitable for assays.
  • White to off-white solid stable under recommended storage.
  • Sequence-defined cyclic peptide for integrin targeting studies.
  • Available in small-scale packaging for laboratory use.
Catalog No. 50-002-12790 Supplier Medchemexpress LLC Supplier No. HY10056325MG
Cyclo(RGDyK) trifluoroacetate is a cyclic pentapeptide integrin inhibitor supplied as the trifluoroacetate salt. It selectively binds the αVβ3 integrin (IC50 ≈ 20 nM) and is provided as a high-purity white to off-white solid for biochemical and cell-based research. For research use only; not for human or clinical use.
  • Selective αVβ3 integrin inhibition with low-nanomolar potency.
  • High chemical purity (~99.9%) suitable for assays.
  • White to off-white solid stable under recommended storage.
  • Sequence-defined cyclic peptide for integrin targeting studies.
  • Available in small-scale packaging for laboratory use.