Learn More
Medchemexpress LLC Cyclo(Arg-Pro) | 74838-83-8 | 98.1% | 253.30 | 5 MG
SDP
Catalog No. 5000426272
Encompass_Preferred
Cyclo(Arg-Pro) (Cyclo(Pro-Arg)) is an inhibitor for chitinase. This compound inhibits cell separation in *Saccharomyces cerevisiae* without affecting its growth, and it also inhibits the morphological change of *Candida albicans* from its yeast form to its filamentous form.
  • Acts as a chitinase inhibitor
  • Inhibits cell separation of *Saccharomyces cerevisiae*
  • Inhibits morphological transition of *Candida albicans* from yeast to filamentous form
Catalog No. 50-004-26272 Supplier Medchemexpress LLC Supplier No. HYP103045MG
Cyclo(Arg-Pro) (Cyclo(Pro-Arg)) is an inhibitor for chitinase. This compound inhibits cell separation in *Saccharomyces cerevisiae* without affecting its growth, and it also inhibits the morphological change of *Candida albicans* from its yeast form to its filamentous form.
  • Acts as a chitinase inhibitor
  • Inhibits cell separation of *Saccharomyces cerevisiae*
  • Inhibits morphological transition of *Candida albicans* from yeast to filamentous form