Learn More
Medchemexpress LLC Cyclo(Arg-Pro) TFA | 98.6% | 367.32 | 10 MG
SDP
Catalog No. 5000425465
Encompass_Preferred
Cyclo(Arg-Pro) TFA is a chitinase inhibitor that disrupts cell separation and morphological transition of yeast by inhibiting chitinase activity. It prevents cell separation of Saccharomyces cerevisiae, leading to the formation of grape-like cell clusters without inhibiting cell growth. Additionally, it blocks the morphological transition of Candida albicans from yeast form to hyphal form, also without inhibiting cell growth.
  • Chitinase inhibitor.
  • Disrupts cell separation and morphological transition of yeast.
  • Prevents cell separation of Saccharomyces cerevisiae without inhibiting cell growth.
  • Blocks morphological transition of Candida albicans from yeast to hyphal form without inhibiting cell growth.
  • For research use only.
Catalog No. 50-004-25465 Supplier Medchemexpress LLC Supplier No. HYP10304A10MG
Cyclo(Arg-Pro) TFA is a chitinase inhibitor that disrupts cell separation and morphological transition of yeast by inhibiting chitinase activity. It prevents cell separation of Saccharomyces cerevisiae, leading to the formation of grape-like cell clusters without inhibiting cell growth. Additionally, it blocks the morphological transition of Candida albicans from yeast form to hyphal form, also without inhibiting cell growth.
  • Chitinase inhibitor.
  • Disrupts cell separation and morphological transition of yeast.
  • Prevents cell separation of Saccharomyces cerevisiae without inhibiting cell growth.
  • Blocks morphological transition of Candida albicans from yeast to hyphal form without inhibiting cell growth.
  • For research use only.