Learn More
Medchemexpress LLC Cyclo(CLLFVY) | 1446322-73-1 | 99.37% | 738.94 | 1 MG
SDP
Catalog No. 5000423752
Encompass_Preferred
cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), with IC50 values of 19 μM (in U2OS) and 16 μM (in MCF-7). It binds to the PAS-B domain of HIF-1α, inhibiting HIF-1 dimerization and transcriptional activity.
  • Downregulates the expression of hypoxia response genes, such as VEGF and CAIX.
  • Exhibits antitumor activity against HIF-1 associated cancers.
Catalog No. 50-004-23752 Supplier Medchemexpress LLC Supplier No. HYP104261MG
cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), with IC50 values of 19 μM (in U2OS) and 16 μM (in MCF-7). It binds to the PAS-B domain of HIF-1α, inhibiting HIF-1 dimerization and transcriptional activity.
  • Downregulates the expression of hypoxia response genes, such as VEGF and CAIX.
  • Exhibits antitumor activity against HIF-1 associated cancers.