Learn More
Medchemexpress LLC Cyclo(CLLFVY) | 1446322-73-1 | 99.37% | 738.94 | 10 MG
SDP
Catalog No. 5000423823
Encompass_Preferred
cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), with IC50 values of 19 μM (in U2OS) and 16 μM (in MCF-7). It binds to the PAS-B domain of HIF-1α, thereby inhibiting HIF-1 dimerization and transcriptional activity. This compound also downregulates the expression of hypoxia response genes, such as VEGF and CAIX, demonstrating antitumor properties against HIF-1 associated cancers.
  • Inhibitor for hypoxia inducible factor-1 (HIF-1)
  • Binds to the PAS-B domain of HIF-1α
  • Inhibits HIF-1 dimerization and transcriptional activity
  • Downregulates the expression of hypoxia response genes (e.g., VEGF and CAIX)
  • Exhibits antitumor activity against HIF-1 associated cancers
Catalog No. 50-004-23823 Supplier Medchemexpress LLC Supplier No. HYP1042610MG
cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), with IC50 values of 19 μM (in U2OS) and 16 μM (in MCF-7). It binds to the PAS-B domain of HIF-1α, thereby inhibiting HIF-1 dimerization and transcriptional activity. This compound also downregulates the expression of hypoxia response genes, such as VEGF and CAIX, demonstrating antitumor properties against HIF-1 associated cancers.
  • Inhibitor for hypoxia inducible factor-1 (HIF-1)
  • Binds to the PAS-B domain of HIF-1α
  • Inhibits HIF-1 dimerization and transcriptional activity
  • Downregulates the expression of hypoxia response genes (e.g., VEGF and CAIX)
  • Exhibits antitumor activity against HIF-1 associated cancers