Learn More
Bachem BQ-123
SDP
Supplier:  Bachem 4016690.0001
Encompass_Preferred
BQ-123 , Cyclo(-D-Trp-D-Asp-Pro-D-Val-Leu), 136553-81-6, C31H42N6O7, Mr 610.71, 1mg. BQ-123 is a potent endothelin antagonist with high selectivity for the ET-A receptor subtype.
Catalog No. 50-259-479