Learn More
AdipoGen trans-4,5-Dihydroxy-1,2-dithiane (5 g)
SDP
Supplier:  AdipoGen CDXD0277G005
Encompass_Preferred
Chemical. CAS: 14193-38-5. Formula: C4H8O2S2. MW: 152.24. trans-4,5-Dihydroxy-1,2-dithiane (oxidized dtt) is an intramolecular disulfide form of dithiothreitol. Oxidized DTT exhibits a strong absorbance peak at 280 nm. It is a strong reducing agent with an ability to elicit a molecular stress response by activating the stress-responsive genes.
Catalog No. 50-205-4378