Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(His-Phe) (TFA) | 147600-13-3 | 99.9% | 398.34 | 25 MG
SDP

Catalog No. 5000400339
Encompass_Preferred

Cyclo(His-Phe) (TFA) is a chemical compound with a molecular weight of 398.34 and a CAS number of 147600-13-3. This product is characterized by its high purity of 99.9% (HPLC) and specific storage requirements to maintain stability.

  • High purity 99.9%
  • Requires sealed storage, away from moisture
  • Powder stable for 2 years at -80°C
  • Powder stable for 1 year at -20°C
  • In solvent, stable for 6 months at -80°C
  • In solvent, stable for 1 month at -20°C
  • Intended for research use only

Catalog No. 50-004-00339 Supplier Medchemexpress LLC Supplier No. 25MGHYP5028A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(His-Phe) (TFA) is a chemical compound with a molecular weight of 398.34 and a CAS number of 147600-13-3. This product is characterized by its high purity of 99.9% (HPLC) and specific storage requirements to maintain stability.

  • High purity 99.9%
  • Requires sealed storage, away from moisture
  • Powder stable for 2 years at -80°C
  • Powder stable for 1 year at -20°C
  • In solvent, stable for 6 months at -80°C
  • In solvent, stable for 1 month at -20°C
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.