Learn More
Medchemexpress LLC Cyclo(histidyl-proline) | 53109-32-3 | MFCD00133207 | 98.8% | 234.25 g/mol | C11H14N4O2 | 1 ML
SDP
Catalog No. 5000211144
Encompass_Preferred
Cyclo(histidyl-proline) is a cyclic dipeptide used in biochemical and cellular research to probe signaling pathways, including NF-κB-related inflammatory responses. The compound is provided as a ready-to-use 10 mM solution in water (1 mL) and is characterized by CAS 53109-32-3, formula C11H14N4O2, and molecular weight 234.25 g/mol.
  • Ready-to-use 10 mM aqueous solution for immediate experiments.
  • High purity for reproducible biochemical assays.
  • Soluble in water and compatible with DMSO for assay flexibility.
  • Well-characterized molecular identity for accurate compound tracking.
Catalog No. 50-002-11144 Supplier Medchemexpress LLC Supplier No. HY10140210MM 1ML
Cyclo(histidyl-proline) is a cyclic dipeptide used in biochemical and cellular research to probe signaling pathways, including NF-κB-related inflammatory responses. The compound is provided as a ready-to-use 10 mM solution in water (1 mL) and is characterized by CAS 53109-32-3, formula C11H14N4O2, and molecular weight 234.25 g/mol.
  • Ready-to-use 10 mM aqueous solution for immediate experiments.
  • High purity for reproducible biochemical assays.
  • Soluble in water and compatible with DMSO for assay flexibility.
  • Well-characterized molecular identity for accurate compound tracking.