Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(histidyl-proline) trifluoroacetate | 936749-56-3 | 98.3% | 348.28 | C13H15F3N4O4 | 1 ML
SDP

Catalog No. 5000247612
Encompass_Preferred

Cyclo(histidyl-proline) trifluoroacetate is the trifluoroacetate salt of the cyclic dipeptide cyclo(His-Pro). It is supplied as a 10 mM solution in DMSO or as solid material and is used in cell-based studies to probe NF-κB signaling, inflammatory responses, and stress-related pathways due to its ability to cross the blood-brain barrier.

  • Supplied as a 10 mM solution in DMSO and as solids.
  • Molecular formula C13H15F3N4O4 and molecular weight 348.28.
  • Reported purity approximately 98.3% (batch-dependent).
  • Soluble in DMSO and water for biological assays.
  • Inhibits NF-κB nuclear accumulation in cell models.
  • Capable of crossing the blood-brain barrier for CNS studies.

Catalog No. 50-002-47612 Supplier Medchemexpress LLC Supplier No. HY101402A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(histidyl-proline) trifluoroacetate is the trifluoroacetate salt of the cyclic dipeptide cyclo(His-Pro). It is supplied as a 10 mM solution in DMSO or as solid material and is used in cell-based studies to probe NF-κB signaling, inflammatory responses, and stress-related pathways due to its ability to cross the blood-brain barrier.

  • Supplied as a 10 mM solution in DMSO and as solids.
  • Molecular formula C13H15F3N4O4 and molecular weight 348.28.
  • Reported purity approximately 98.3% (batch-dependent).
  • Soluble in DMSO and water for biological assays.
  • Inhibits NF-κB nuclear accumulation in cell models.
  • Capable of crossing the blood-brain barrier for CNS studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.