Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(Ile-Ala) | 90821-99-1 | 98.5% | 184.24 | 10 MG
SDP

Catalog No. 5000426713
Encompass_Preferred

Cyclo(Ile-Ala) is a cyclic dipeptide found in marine actinomycete 11014 I. It is intended for research use only.

  • Purity of 98.47%
  • Appears as a solid
  • White to off-white color
  • Molecular weight of 184.24
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-26713 Supplier Medchemexpress LLC Supplier No. HYN925110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(Ile-Ala) is a cyclic dipeptide found in marine actinomycete 11014 I. It is intended for research use only.

  • Purity of 98.47%
  • Appears as a solid
  • White to off-white color
  • Molecular weight of 184.24
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.