Learn More
Medchemexpress LLC Cyclo(L-arginylglycyl-L-a-aspartyl-D-phenylalanyl-N-methyl-L-valyl) | 188968-51-6 | 99.8% | 588.66 | 200 MG
SDP
Catalog No. 5000391491
Encompass_Preferred
Cilengitide (HY-16141) is a blood-brain barrier-permeable integrin antagonist. It inhibits the binding of ανβ3 and ανβ5 to Vitronectin and also inhibits TGF-β/Smad signaling, mediating PD-L1 expression. Cilengitide induces apoptosis and exhibits an antiangiogenic effect in research against glioblastoma and other cancers.
  • BΒB-permeable integrin antagonist.
  • Inhibits binding to Vitronectin.
  • Inhibits TGF-β/Smad signaling.
  • Mediates PD-L1 expression.
  • Induces apoptosis.
  • Shows antiangiogenic effect.
Catalog No. 50-003-91491 Supplier Medchemexpress LLC Supplier No. HY16141200MG
Cilengitide (HY-16141) is a blood-brain barrier-permeable integrin antagonist. It inhibits the binding of ανβ3 and ανβ5 to Vitronectin and also inhibits TGF-β/Smad signaling, mediating PD-L1 expression. Cilengitide induces apoptosis and exhibits an antiangiogenic effect in research against glioblastoma and other cancers.
  • BΒB-permeable integrin antagonist.
  • Inhibits binding to Vitronectin.
  • Inhibits TGF-β/Smad signaling.
  • Mediates PD-L1 expression.
  • Induces apoptosis.
  • Shows antiangiogenic effect.