Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(L-cysteinyl-L-leucyl-L-leucyl-L-phenylalanyl-L-valyl-L-tyrosyl) | 1446322-73-1 | 99.37% | 738.94 | 5 MG
SDP

Catalog No. 5000423824
Encompass_Preferred

Cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), demonstrating IC50 values of 19 μM in U2OS cells and 16 μM in MCF-7 cells. This compound functions by binding to the PAS-B domain of HIF-1α, thereby inhibiting HIF-1 dimerization and its transcriptional activity. It effectively downregulates the expression of hypoxia response genes, including VEGF and CAIX, and has shown antitumor effects against HIF-1 associated cancers.

  • Inhibitor for hypoxia inducible factor-1 (HIF-1)
  • Binds to the PAS-B domain of HIF-1α
  • Inhibits HIF-1 dimerization and transcriptional activity
  • Downregulates expression of hypoxia response genes (VEGF, CAIX)
  • Exhibits antitumor effects against HIF-1 associated cancers
  • Appearance: White to off-white solid
  • Solubility: 100 mg/mL in DMSO
  • Long-term storage: -80°C for powder

Catalog No. 50-004-23824 Supplier Medchemexpress LLC Supplier No. HYP104265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(CLLFVY) is an inhibitor for hypoxia inducible factor-1 (HIF-1), demonstrating IC50 values of 19 μM in U2OS cells and 16 μM in MCF-7 cells. This compound functions by binding to the PAS-B domain of HIF-1α, thereby inhibiting HIF-1 dimerization and its transcriptional activity. It effectively downregulates the expression of hypoxia response genes, including VEGF and CAIX, and has shown antitumor effects against HIF-1 associated cancers.

  • Inhibitor for hypoxia inducible factor-1 (HIF-1)
  • Binds to the PAS-B domain of HIF-1α
  • Inhibits HIF-1 dimerization and transcriptional activity
  • Downregulates expression of hypoxia response genes (VEGF, CAIX)
  • Exhibits antitumor effects against HIF-1 associated cancers
  • Appearance: White to off-white solid
  • Solubility: 100 mg/mL in DMSO
  • Long-term storage: -80°C for powder

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.