Learn More
Cayman Chemical Cyclo(L-His-L-Pro), Purity: ≥95%, CAS: 53109-32-3, 50 mg
SDP
Catalog No. 502989511
Encompass_Preferred
A cyclic neuropeptide and metabolite of TRH; ubiquitously expressed in the CNS, but is also found in the gastrointestinal tract and blood; reduces LPS-induced production of ROS and NO, as well as translocation of NF-κB in BV-2 microglia; levels are increased in rat brain after six weeks of continuous ethanol consumption; reduces ethanol-induced sleep time in rats at 1 µmol/kg; induces analgesia in the hot-plate test and reduces acetic acid-induced writhing in mice
Catalog No. 50-298-9511 Supplier Cayman Chemical Supplier No. 3117250 MG
A cyclic neuropeptide and metabolite of TRH; ubiquitously expressed in the CNS, but is also found in the gastrointestinal tract and blood; reduces LPS-induced production of ROS and NO, as well as translocation of NF-κB in BV-2 microglia; levels are increased in rat brain after six weeks of continuous ethanol consumption; reduces ethanol-induced sleep time in rats at 1 µmol/kg; induces analgesia in the hot-plate test and reduces acetic acid-induced writhing in mice