Learn More
Cayman Chemical Cyclo(L-Leu-L-Pro), Purity: ≥95%, CAS: 2873-36-1, 5 mg
SDP
Catalog No. 502934045
Encompass_Preferred
A diketopiperazine metabolite; active against VRE strains of E. faecalis including K-99-34, K-00-184, and K-00-221 (MICs = 12.5 µg/ml); inhibits growth of K562, HL-60, and U937 leukemia cells (1-100 µg/ml); has antifouling activity, inhibiting attachment of B. amphitrite larva (EC50 = 0.15 mM)
Catalog No. 50-293-4045 Supplier Cayman Chemical Supplier No. 249415 MG
A diketopiperazine metabolite; active against VRE strains of E. faecalis including K-99-34, K-00-184, and K-00-221 (MICs = 12.5 µg/ml); inhibits growth of K562, HL-60, and U937 leukemia cells (1-100 µg/ml); has antifouling activity, inhibiting attachment of B. amphitrite larva (EC50 = 0.15 mM)