Learn More
Cayman Chemical Cyclo(L-Leu-L-Trp), Purity: ≥95%, CAS: 15136-34-2, 5 mg
SDP
Catalog No. 502934242
Encompass_Preferred
A diketopiperazine metabolite; active against various bacteria (MICs = 125-1000 µg/ml) and fungi (MICs = 8-64 µg/ml); inhibits hydroxy radical production (IC50 = 1.8 µM); a bitter tastant that can rapidly permeate rat taste cell membranes (1 mM); a melatonin receptor agonist, inhibiting cAMP accumulation (20 µM)
Catalog No. 50-293-4242 Supplier Cayman Chemical Supplier No. 249425 MG
A diketopiperazine metabolite; active against various bacteria (MICs = 125-1000 µg/ml) and fungi (MICs = 8-64 µg/ml); inhibits hydroxy radical production (IC50 = 1.8 µM); a bitter tastant that can rapidly permeate rat taste cell membranes (1 mM); a melatonin receptor agonist, inhibiting cAMP accumulation (20 µM)