Learn More
Medchemexpress LLC Cyclo-(L-phenylalanyl-4R-hydroxy-L-proline) | 118477-06-8 | 99.8% | 260.29 | C14H16N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1251310MG
Encompass_Preferred
Cyclo(L-Phe-trans-4-OH-L-Pro) is a cyclic dipeptide (antifungal peptide) supplied for research applications. It is provided as a high-purity white to off-white powder intended for biochemical and analytical use. Storage recommendations include long-term storage at -20 °C in the solid form to preserve stability.
  • antifungal cyclic dipeptide for research applications.
  • high purity suitable for analytical and biochemical assays.
  • available in small research pack sizes, including 10 mg.
  • powder stable at -20 °C with multiyear shelf life.
  • for research use only; not for human or clinical use.
Catalog No. 50-002-96580