Learn More
Medchemexpress LLC Cyclo(L-Trp-L-Trp) | 20829-55-4 | 99.8% | 372.42 | 100 MG
SDP
Catalog No. 5000422892
Encompass_Preferred
Cyclo(L-Trp-L-Trp) is an antibiotic that exhibits antimicrobial activity. It is effective against *A. baumannii*, *Candida albicans*, *Bacillus subtilis*, *Micrococcus luteus*, *Saccharomyces cerevisiae*, *Aspergillus niger*, and *Staphylococcus aureus*. It can be utilized in microbial infection research.
  • Antibiotic with antimicrobial activity.
  • Inhibits a range of microorganisms, including bacteria and fungi.
  • Suitable for microbial infection research.
Catalog No. 50-004-22892 Supplier Medchemexpress LLC Supplier No. HYP2124100MG
Cyclo(L-Trp-L-Trp) is an antibiotic that exhibits antimicrobial activity. It is effective against *A. baumannii*, *Candida albicans*, *Bacillus subtilis*, *Micrococcus luteus*, *Saccharomyces cerevisiae*, *Aspergillus niger*, and *Staphylococcus aureus*. It can be utilized in microbial infection research.
  • Antibiotic with antimicrobial activity.
  • Inhibits a range of microorganisms, including bacteria and fungi.
  • Suitable for microbial infection research.