Learn More
Cayman Chemical Cyclo(L-Trp-L-Trp), Purity: ≥98%, CAS: 20829-55-4, 50 mg
SDP
Catalog No. 502995293
Encompass_Preferred
A cyclic dipeptide; used as a substrate for indole prenyltransferases in the synthesis of mono- and diprenylated indolines; inhibits growth of 41 out of 49 strains of multidrug resistant A. baumannii (MICs = 12.5-25 µg/ml) as well as B. subtilis, M. luteus, S. aureus, S. cerevisiae, A. niger, and C. albicans (MICs = 12.5-50 µg/ml)
Catalog No. 50-299-5293 Supplier Cayman Chemical Supplier No. 2349450 MG
A cyclic dipeptide; used as a substrate for indole prenyltransferases in the synthesis of mono- and diprenylated indolines; inhibits growth of 41 out of 49 strains of multidrug resistant A. baumannii (MICs = 12.5-25 µg/ml) as well as B. subtilis, M. luteus, S. aureus, S. cerevisiae, A. niger, and C. albicans (MICs = 12.5-50 µg/ml)