Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(-Leu-Phe) | 7280-77-5 | 99.6% | 260.33 | 10 MG
SDP

Catalog No. 5000397512
Encompass_Preferred

Cyclo(-Leu-Phe) is a cyclic peptide composed of leucine and phenylalanine, forming a ring structure through peptide bonds. It is for research use only.

  • Composed of leucine and phenylalanine
  • Forms a ring structure through peptide bonds
  • White to off-white solid
  • Soluble in DMSO (4.6 mg/mL)
  • For research use only
  • Recommended storage is sealed and away from moisture
  • Powder is stable for 2 years at -80°C and 1 year at -20°C
  • In solvent, it is stable for 6 months at -80°C and 1 month at -20°C

Catalog No. 50-003-97512 Supplier Medchemexpress LLC Supplier No. HYP461710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(-Leu-Phe) is a cyclic peptide composed of leucine and phenylalanine, forming a ring structure through peptide bonds. It is for research use only.

  • Composed of leucine and phenylalanine
  • Forms a ring structure through peptide bonds
  • White to off-white solid
  • Soluble in DMSO (4.6 mg/mL)
  • For research use only
  • Recommended storage is sealed and away from moisture
  • Powder is stable for 2 years at -80°C and 1 year at -20°C
  • In solvent, it is stable for 6 months at -80°C and 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.