Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(-Leu-Phe) | 7280-77-5 | 99.6% | 260.33 | 100 MG
SDP

Catalog No. 5000397507
Encompass_Preferred

Cyclo(-Leu-Phe) is a cyclic peptide composed of leucine and phenylalanine, forming a ring structure through peptide bonds.

  • Molecular formula: C15H20N2O2
  • CAS number: 7280-77-5
  • Sequence: Cyclo(-Leu-Phe)
  • Sequence shortening: Cyclo(-LF)
  • Purity: 99.58%
  • Storage (powder): Sealed, away from moisture, 2 years at -80°C, 1 year at -20°C
  • Storage (in solvent): Sealed, away from moisture, 6 months at -80°C, 1 month at -20°C
  • Solubility (in vitro): 4.6 mg/mL (17.67 mM) in DMSO; requires ultrasonic and warming

Catalog No. 50-003-97507 Supplier Medchemexpress LLC Supplier No. HYP4617100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(-Leu-Phe) is a cyclic peptide composed of leucine and phenylalanine, forming a ring structure through peptide bonds.

  • Molecular formula: C15H20N2O2
  • CAS number: 7280-77-5
  • Sequence: Cyclo(-Leu-Phe)
  • Sequence shortening: Cyclo(-LF)
  • Purity: 99.58%
  • Storage (powder): Sealed, away from moisture, 2 years at -80°C, 1 year at -20°C
  • Storage (in solvent): Sealed, away from moisture, 6 months at -80°C, 1 month at -20°C
  • Solubility (in vitro): 4.6 mg/mL (17.67 mM) in DMSO; requires ultrasonic and warming

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.