Learn More
Bachem Cyclo(-Leu-Pro)
SDP
Supplier:  Bachem 4003115.0250
Encompass_Preferred
Cyclo(-Leu-Pro), 2873-36-1, C11H18N2O2, Mr 210.28, 250mg. Cyclo(Leu-Pro) showed antibacterial activity. Yan et al. observed that Cyclo(Leu-Pro) produced by Achromobacter xylosoxidans inhibited aflatoxin production by Aspergillus parasiticus. The diketopiperazine was shown to inhibit catecholamine release in vitro. It could play a role in the hypothalamic modulation of appetite suppressant circuitry.
Catalog No. 50-259-286