Learn More
Medchemexpress LLC Cyclo(Phe-Phe) | 2862-51-3 | MFCD00190985 | 98.8% | 294.35 | C18H18N2O2 | 25 MG
SDP
Catalog No. 5000283998
Encompass_Preferred
Cyclo(Phe-Phe) is a cyclic dipeptide consisting of two phenylalanine residues. Supplied as a white to off-white solid for research use, it is employed in peptide screening, protein interaction studies, functional analysis, and epitope mapping. Identifiers: CAS 2862-51-3; molecular formula C18H18N2O2; molecular weight 294.35; reported purity 98.8%.
  • High purity (98.8%).
  • Defined molecular formula and molecular weight (C18H18N2O2; 294.35).
  • Soluble in DMSO with ultrasonic warming; heating to 60°C improves solubility.
  • Suitable for peptide screening, protein interaction, and epitope mapping assays.
  • Available in small sizes (25 mg) with larger quantities on request.
  • Stable when stored sealed at -80°C or -20°C under recommended conditions.
Catalog No. 50-002-83998 Supplier Medchemexpress LLC Supplier No. HYW54943825MG
Cyclo(Phe-Phe) is a cyclic dipeptide consisting of two phenylalanine residues. Supplied as a white to off-white solid for research use, it is employed in peptide screening, protein interaction studies, functional analysis, and epitope mapping. Identifiers: CAS 2862-51-3; molecular formula C18H18N2O2; molecular weight 294.35; reported purity 98.8%.
  • High purity (98.8%).
  • Defined molecular formula and molecular weight (C18H18N2O2; 294.35).
  • Soluble in DMSO with ultrasonic warming; heating to 60°C improves solubility.
  • Suitable for peptide screening, protein interaction, and epitope mapping assays.
  • Available in small sizes (25 mg) with larger quantities on request.
  • Stable when stored sealed at -80°C or -20°C under recommended conditions.