Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(phe-phe) | 2862-51-3 | MFCD00190985 | 98.8% | 294.35 g/mol | C18H18N2O2 | 50 MG
SDP

Catalog No. 5000284210
Encompass_Preferred

Cyclo(Phe-Phe) is a cyclic dipeptide (cyclo-phenylalanyl-phenylalanine) used as a research reagent in peptide screening and biochemical assays. It is supplied as a white to off-white solid with high purity; handle according to the safety data sheet guidance.

  • High purity: 98.8%.
  • Molecular weight: 294.35 g/mol.
  • Chemical formula: C18H18N2O2.
  • Physical state: Solid, white to off-white.
  • Recommended storage: Powder at -80°C (up to 2 years) or -20°C (up to 1 year), sealed.
  • Documentation: Datasheet, certificate of analysis, and SDS available.

Catalog No. 50-002-84210 Supplier Medchemexpress LLC Supplier No. HYW54943850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(Phe-Phe) is a cyclic dipeptide (cyclo-phenylalanyl-phenylalanine) used as a research reagent in peptide screening and biochemical assays. It is supplied as a white to off-white solid with high purity; handle according to the safety data sheet guidance.

  • High purity: 98.8%.
  • Molecular weight: 294.35 g/mol.
  • Chemical formula: C18H18N2O2.
  • Physical state: Solid, white to off-white.
  • Recommended storage: Powder at -80°C (up to 2 years) or -20°C (up to 1 year), sealed.
  • Documentation: Datasheet, certificate of analysis, and SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.