Learn More
Bachem Cyclo(-Phe-Pro)
SDP
Supplier:  Bachem 4029456.1000
Encompass_Preferred
Cyclo(-Phe-Pro), 3705-26-8, C14H16N2O2, Mr 244.29, 1g. Cain et al. obtained an array of small molecule peptide mimetics targeting the melanocortin receptors from this diketopiperazine. Brauns et al. demonstrated that cyclo(Phe-Pro) inhibits the growth of HT-29, MCF-7 and HeLa cells and induces apoptosis in HT-29 colon cancer cells.
Catalog No. 50-259-449