Learn More
Cayman Chemical Cyclo Phe-Pro 100mg
SDP
Catalog No. 502910565
Encompass_Preferred
A bacterial quorum-sensing molecule; inhibits virulence factor production in V. cholerae; inhibitor of the RIG-I-TRIM25 interaction; increases the viral load in Huh7 cells infected with HCV at 1, 2.5, and 5 mM; reduces the expression of IFN-β induced by poly(I:C) and increases serum HCV titers in animal models of HCV infection
Catalog No. 50-291-0565 Supplier Cayman Chemical Supplier No. 34758100 MG
A bacterial quorum-sensing molecule; inhibits virulence factor production in V. cholerae; inhibitor of the RIG-I-TRIM25 interaction; increases the viral load in Huh7 cells infected with HCV at 1, 2.5, and 5 mM; reduces the expression of IFN-β induced by poly(I:C) and increases serum HCV titers in animal models of HCV infection