Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo-(Pro-gly) (pyrrolopiperazine-2,5-dione) | 19179-12-5 | MFCD01083707 | >98.0% | 154.17 g/mol | C7H10N2O2 | 250 MG
SDP

Catalog No. 5000209412
Encompass_Preferred

Cyclo-(Pro-Gly) is a cyclic dipeptide (diketopiperazine) used as an analytical standard and reference compound for research applications. It is employed in analytical method development and studies of small natural-product-derived peptides.

  • Used as an analytical standard for method development
  • Molecular weight 154.17 g/mol
  • CAS number 19179-12-5
  • Chemical formula C7H10N2O2
  • Suitable for research and analytical reference purposes

Catalog No. 50-002-09412 Supplier Medchemexpress LLC Supplier No. 250MGHYW062171
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo-(Pro-Gly) is a cyclic dipeptide (diketopiperazine) used as an analytical standard and reference compound for research applications. It is employed in analytical method development and studies of small natural-product-derived peptides.

  • Used as an analytical standard for method development
  • Molecular weight 154.17 g/mol
  • CAS number 19179-12-5
  • Chemical formula C7H10N2O2
  • Suitable for research and analytical reference purposes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.