Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo-(Pro-Gly) (pyrrolopiperazine-2,5-dione) | 19179-12-5 | MFCD01083707 | 99.9% | 154.17 g/mol | C7H10N2O2 | 1 G
SDP

Catalog No. 5000210505
Encompass_Preferred

Cyclo-(Pro-Gly) is a cyclic dipeptide (pyrrolopiperazine-2,5-dione) used as a reference standard in biochemical and pharmacological research. It is supplied as a high-purity solid and as a 10 mM solution in DMSO for laboratory assays; intended for research use only.

  • High purity: 99.9% (HPLC).
  • Chemical name: Cyclo-(Pro-Gly) (pyrrolopiperazine-2,5-dione).
  • Cas number: 19179-12-5.
  • Molecular formula: C7H10N2O2.
  • Molecular weight: 154.17 g/mol.
  • Available formats: solid (50 mg-10 g) and 10 mM solution in DMSO (1 mL).
  • Intended use: research only; not for human or clinical use.
  • Storage: follow conditions in the certificate of analysis.

Catalog No. 50-002-10505 Supplier Medchemexpress LLC Supplier No. HYW0621711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo-(Pro-Gly) is a cyclic dipeptide (pyrrolopiperazine-2,5-dione) used as a reference standard in biochemical and pharmacological research. It is supplied as a high-purity solid and as a 10 mM solution in DMSO for laboratory assays; intended for research use only.

  • High purity: 99.9% (HPLC).
  • Chemical name: Cyclo-(Pro-Gly) (pyrrolopiperazine-2,5-dione).
  • Cas number: 19179-12-5.
  • Molecular formula: C7H10N2O2.
  • Molecular weight: 154.17 g/mol.
  • Available formats: solid (50 mg-10 g) and 10 mM solution in DMSO (1 mL).
  • Intended use: research only; not for human or clinical use.
  • Storage: follow conditions in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.