Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(Pro-Thr) | 227777-31-3 | MFCD05663455 | 98.15% | 198.22 | 5 MG
SDP

Catalog No. 5000398422
Encompass_Preferred

Cyclo(Pro-Thr) is a polypeptide that can be identified through peptide screening, a research tool used for protein interaction, functional analysis, and epitope screening in agent research and development. It has a molecular weight of 198.22, a chemical formula of C9H14N2O3, and appears as a white to off-white solid. This compound has been reported in *Streptomyces nigra*.

  • High purity of 98.15%
  • Molecular weight of 198.22
  • Chemical formula C9H14N2O3
  • CAS number 227777-31-3
  • Solid appearance, white to off-white color
  • Peptide sequence Cyclo(Pro-Thr)
  • MFCD number MFCD05663455
  • Utilized in peptide screening for protein interaction and functional analysis
  • Reported in *Streptomyces nigra*

Catalog No. 50-003-98422 Supplier Medchemexpress LLC Supplier No. HYP50505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(Pro-Thr) is a polypeptide that can be identified through peptide screening, a research tool used for protein interaction, functional analysis, and epitope screening in agent research and development. It has a molecular weight of 198.22, a chemical formula of C9H14N2O3, and appears as a white to off-white solid. This compound has been reported in *Streptomyces nigra*.

  • High purity of 98.15%
  • Molecular weight of 198.22
  • Chemical formula C9H14N2O3
  • CAS number 227777-31-3
  • Solid appearance, white to off-white color
  • Peptide sequence Cyclo(Pro-Thr)
  • MFCD number MFCD05663455
  • Utilized in peptide screening for protein interaction and functional analysis
  • Reported in *Streptomyces nigra*

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.