Learn More
Bachem Cyclo(-Pro-Tyr)
SDP
Supplier:  Bachem 4029436.0250
Encompass_Preferred
Cyclo(-Pro-Tyr), 4549-02-4, C14H16N2O3, Mr 260.29, 250mg. Maculosin. The host-specific phytotoxin cyclo(Pro-Tyr) produced by Alternaria alternata on spotted knapweed is an ideal prototype for creating a safe and an environmentally friendly antiknapweed herbicide. Furthermore, this cyclic dipeptide induced a high degree of phosphorylation of histones.
Catalog No. 50-382-351