Learn More
Enzo Life Sciences Cyclo [Arg-Gly-Asp-D-Phe-Val] (1mg). CAS: 137813-35-5
SDP
Catalog No. 502010303
Encompass_Preferred
Integrin αvβ3 antagonist. Inhibits cell adhesion events mediated by αvβ3. Inhibits angiogenesis. Induces rapid regression of histologically distinct human tumors. Induces apoptosis of angiogenic blood vessels. Purity: ≥98%. Solubility: Soluble in DMSO (5 mg/ml) and PBS (1 mg/350μl). Long Term Storage: -20°C.
Catalog No. 50-201-0303 Supplier Enzo Life Sciences Supplier No. BMLAM1000001
Integrin αvβ3 antagonist. Inhibits cell adhesion events mediated by αvβ3. Inhibits angiogenesis. Induces rapid regression of histologically distinct human tumors. Induces apoptosis of angiogenic blood vessels. Purity: ≥98%. Solubility: Soluble in DMSO (5 mg/ml) and PBS (1 mg/350μl). Long Term Storage: -20°C.