Learn More
Cayman Chemical Cyclo(RGDfC) (trifluoroacetate salt), Purity: ≥90%, 10 mg
SDP
Catalog No. 502910925
Encompass_Preferred
A cyclic pentapeptide; has been used in the synthesis of unimolecular micelles and ligand-decorated nanogels for cancer cell-targeted drug delivery
Catalog No. 50-291-0925 Supplier Cayman Chemical Supplier No. 3517210 MG
A cyclic pentapeptide; has been used in the synthesis of unimolecular micelles and ligand-decorated nanogels for cancer cell-targeted drug delivery