Learn More
Cayman Chemical Cyclo(RGDfK), Purity: ≥95%, CAS: 161552-03-0, 100 mg
SDP
Catalog No. 502990096
Encompass_Preferred
An inhibitor of αvβ3 integrin (IC50 = 1.33 nM); taken up into tumors in vivo and in vitro and it has been polymerized or conjugated to various fluorophores, radiolabels, and peptide sequences for use in fluorescent and PET imaging to study tumor cell adhesion
Catalog No. 50-299-0096 Supplier Cayman Chemical Supplier No. 29514100 MG
An inhibitor of αvβ3 integrin (IC50 = 1.33 nM); taken up into tumors in vivo and in vitro and it has been polymerized or conjugated to various fluorophores, radiolabels, and peptide sequences for use in fluorescent and PET imaging to study tumor cell adhesion