Learn More
Apexbio Technology LLC Cyclo (-RGDfK) 161552-03-0 100mg
SDP
Catalog No. 5000062650
Encompass_Preferred
Cyclo(-RGDfK) is a cyclic pentapeptide that functions as an inhibitor targeting the integrin v 3 Integrins are transmembrane receptors mediating cell adhesion by recognizing specific peptide sequences notably the Arg-Gly-Asp (RGD) motif Integrin v 3 is involved in cell adhesion processes such as angiogenesis tissue remodeling and tumor development and is abundantly expressed on activated endothelial cells and various tumor cells Cyclo(-RGDfK) specifically binds integrin v 3 disrupting integrin-mediated cell attachment Consequently it is frequently utilized in bioengineering applications to modify biomaterial surfaces for enhanced cellular adherence and integration Additionally cyclo(-RGDfK) serves as a targeting ligand in imaging probes aimed at localizing integrin-positive tissues in cancer research
Catalog No. 50-000-62650 Supplier Apexbio Technology LLC Supplier No. A8164100
Add to Cart
Add to Cart
Cyclo(-RGDfK) is a cyclic pentapeptide that functions as an inhibitor targeting the integrin v 3 Integrins are transmembrane receptors mediating cell adhesion by recognizing specific peptide sequences notably the Arg-Gly-Asp (RGD) motif Integrin v 3 is involved in cell adhesion processes such as angiogenesis tissue remodeling and tumor development and is abundantly expressed on activated endothelial cells and various tumor cells Cyclo(-RGDfK) specifically binds integrin v 3 disrupting integrin-mediated cell attachment Consequently it is frequently utilized in bioengineering applications to modify biomaterial surfaces for enhanced cellular adherence and integration Additionally cyclo(-RGDfK) serves as a targeting ligand in imaging probes aimed at localizing integrin-positive tissues in cancer research