Learn More
Cayman Chemical Cyclo(RGDfK), Purity: ≥95%, CAS: 161552-03-0, 10 mg
SDP
Catalog No. 502988991
Encompass_Preferred
An inhibitor of αvβ3 integrin (IC50 = 1.33 nM); taken up into tumors in vivo and in vitro and it has been polymerized or conjugated to various fluorophores, radiolabels, and peptide sequences for use in fluorescent and PET imaging to study tumor cell adhesion
Catalog No. 50-298-8991 Supplier Cayman Chemical Supplier No. 2951410 MG
An inhibitor of αvβ3 integrin (IC50 = 1.33 nM); taken up into tumors in vivo and in vitro and it has been polymerized or conjugated to various fluorophores, radiolabels, and peptide sequences for use in fluorescent and PET imaging to study tumor cell adhesion