Learn More
Medchemexpress LLC Cyclo(-RGDfK) | 161552-03-0 | 99.8% | 603.67 | 1 MG
SDP
Catalog No. 5000426494
Encompass_Preferred
Cyclo(-RGDfK) is a potent and selective inhibitor of the αvβ3 integrin, with an IC50 of 0.94 nM. It targets tumor microvasculature and cancer cells by specifically binding to the αvβ3 integrin on the cell surface.
  • Potent and selective inhibitor of αvβ3 integrin
  • Targets tumor microvasculature and cancer cells
Catalog No. 50-004-26494 Supplier Medchemexpress LLC Supplier No. HYP00231MG
Add to Cart
Add to Cart
Cyclo(-RGDfK) is a potent and selective inhibitor of the αvβ3 integrin, with an IC50 of 0.94 nM. It targets tumor microvasculature and cancer cells by specifically binding to the αvβ3 integrin on the cell surface.
  • Potent and selective inhibitor of αvβ3 integrin
  • Targets tumor microvasculature and cancer cells