Learn More
Medchemexpress LLC Cyclo(-RGDfK) | 161552-03-0 | 99.8% | 603.67 | 50 MG
SDP
Catalog No. 5000426512
Encompass_Preferred
Cyclo(-RGDfK) is a potent and selective inhibitor of the αvβ3 integrin, with an IC50 of 0.94 nM. It targets tumor microvasculature and cancer cells by specifically binding to the αvβ3 integrin on the cell surface.
  • Potent and selective inhibitor of αvβ3 integrin
  • IC50 of 0.94 nM for αvβ3
  • Targets tumor microvasculature and cancer cells
  • Binds specifically to αvβ3 integrin on the cell surface
  • White to off-white solid
Catalog No. 50-004-26512 Supplier Medchemexpress LLC Supplier No. HYP002350MG
Add to Cart
Add to Cart
Cyclo(-RGDfK) is a potent and selective inhibitor of the αvβ3 integrin, with an IC50 of 0.94 nM. It targets tumor microvasculature and cancer cells by specifically binding to the αvβ3 integrin on the cell surface.
  • Potent and selective inhibitor of αvβ3 integrin
  • IC50 of 0.94 nM for αvβ3
  • Targets tumor microvasculature and cancer cells
  • Binds specifically to αvβ3 integrin on the cell surface
  • White to off-white solid