Learn More
Apexbio Technology LLC Cyclo (-RGDfK), 5mg. Cas: 161552-03-0 MFCD: MFCD00937400. Inhibitor of αvβ3 integrin
SDP
Catalog No. 501905066
Encompass_Preferred
In one study where this peptide was labeled with 125I, it was found to bind specifically and with high affinity to v3 receptors on neovascular blood vessel sections of different major human cancers. The integrin alpha(IIb)beta(3)-specific cyclic hexapeptide contains an Arg-Gly-Asp (RGD) sequence. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5066 Supplier Apexbio Technology LLC Supplier No. A81645
In one study where this peptide was labeled with 125I, it was found to bind specifically and with high affinity to v3 receptors on neovascular blood vessel sections of different major human cancers. The integrin alpha(IIb)beta(3)-specific cyclic hexapeptide contains an Arg-Gly-Asp (RGD) sequence. Other sizes are available. Please inqury us for quote.