Learn More
Carbosynth LLC. H-dPEG4-Glu[dPEG4(Cyclo(Arg-Gly-Asp-d-Phe-Lys))]2, 5mg, Clickable RGD peptide for Click chemistry., MW: 2060.35, Molecular formula: C92H150N22O31
SDP
Supplier:  Carbosynth LLC. RGD3750PI5MG
Encompass_Preferred
Synonyms: 3P-RGD2, 3PRGD2, PEG4-E[PEG4-c(RGKfK)]2 = PEG4-Glu[cyclo[Arg-Gly-Asp-D-Phe-Lys(PEG4)]]2, H-dPEG4-Glu[dPEG4(cyclo (Arg-Gly-Asp-d-Phe-Lys))]2, Application: For In Vitro Assays of Integrin v3/v5 Expression in Tumor Tissues, Salt Form:, Trifluoroacetate, Storage Temperature: -20 C, References: R. Gao, et al., Med Princ Pract., 24, 244 (2015)., Y Zheng, et al., Bioconjugate Chem, 25,1925 (2014).
Catalog No. 50-168-6935