Learn More
TARGETMOL CHEMICALS INC Cyclo-RGDfK TFA 5MG
SDP
Catalog No. 502879098
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Cyclo(-RGDfK) is a selective alphavbeta3 integrin inhibitor(IC50 0.94 nM).
Catalog No. 50-287-9098 Supplier TARGETMOL CHEMICALS INC Supplier No. T6812L5MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Cyclo(-RGDfK) is a selective alphavbeta3 integrin inhibitor(IC50 0.94 nM).