Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclo(RGDyC) | 1206475-79-7 | 98.3% | 594.64 | 5 MG
SDP

Catalog No. 5000399164
Encompass_Preferred

Cyclo(RGDyC) is a cyclic pentapeptide with anti-angiogenic properties. It can be combined with liposome delivery systems for research related to ocular neovascular diseases and cancer.

  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture and light
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-99164 Supplier Medchemexpress LLC Supplier No. HYP58405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclo(RGDyC) is a cyclic pentapeptide with anti-angiogenic properties. It can be combined with liposome delivery systems for research related to ocular neovascular diseases and cancer.

  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture and light
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.