Learn More
AnaSpec CYCLO(-RGDYK) 5MG
Supplier:  AnaSpec AS611835
Encompass
Cyclo (-RGDyK)[Cyclo(-RGDyK)]; MW 619.7; (5 mg) This is a cyclic derivative of RGD having a high affinity for a vB 3 integrin. It is considered the most prominent structure for the development of molecular imaging compounds in order to determine a vB 3 expression. It has been shown that both purified and membrane-bound a vB 3 intergrin binds to c(RGDyK) with high affinity.
Catalog No. NC0625534