Learn More
Medchemexpress LLC HY-100563 100mg Medchemexpress, Cyclo(RGDyK) trifluoroacetate CAS:250612-42-1 Purity:>98%
SDP
Catalog No. 501931882
Encompass_Preferred
Medchemexpress, HY-100563 100mg Cyclo(RGDyK) trifluoroacetate CAS:250612-42-1 Cyclo(RGDyK) trifluoroacetate is a potent and selective α V β 3 integrin inhibitor with an IC 50 of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1882 Supplier Medchemexpress LLC Supplier No. HY100563100MG
Medchemexpress, HY-100563 100mg Cyclo(RGDyK) trifluoroacetate CAS:250612-42-1 Cyclo(RGDyK) trifluoroacetate is a potent and selective α V β 3 integrin inhibitor with an IC 50 of 20 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.