Learn More
Cayman Chemical Cyclo RGDyKtrIfluoroACTAt 10mg
SDP
Catalog No. 502910427
Encompass_Preferred
A cyclic peptide ligand of αVβ3 integrin (IC50 = 37.5 nM); has been conjugated to various radiolabels or fluorescent probes for the visualization of αVβ3-mediated neoangiogenesis or optical imaging of αVβ3-expressing tumors in live mice; has also been conjugated to SN38 to form a prodrug that targets αVβ3-overexpressing cells and induces cytotoxicity in KB cells
Catalog No. 50-291-0427 Supplier Cayman Chemical Supplier No. 3517310 MG
A cyclic peptide ligand of αVβ3 integrin (IC50 = 37.5 nM); has been conjugated to various radiolabels or fluorescent probes for the visualization of αVβ3-mediated neoangiogenesis or optical imaging of αVβ3-expressing tumors in live mice; has also been conjugated to SN38 to form a prodrug that targets αVβ3-overexpressing cells and induces cytotoxicity in KB cells