Learn More
Medchemexpress LLC Cyclo(RLsKDK) (BK-1361) | 1975145-82-4 | 98.9% | 727.85 | 10 MG
SDP
Catalog No. 5000424008
Encompass_Preferred
Cyclo(RLsKDK) is a specific inhibitor of metalloproteinase ADAM8 with an IC50 value of 182 nM. It has potential applications in inflammatory diseases and cancer. This product is for research use only.
  • Specific inhibitor of metalloproteinase ADAM8
  • IC50 value of 182 nM
  • Potential applications in inflammatory diseases
  • Potential applications in cancer
  • For research use only
Catalog No. 50-004-24008 Supplier Medchemexpress LLC Supplier No. HYP167610MG
Cyclo(RLsKDK) is a specific inhibitor of metalloproteinase ADAM8 with an IC50 value of 182 nM. It has potential applications in inflammatory diseases and cancer. This product is for research use only.
  • Specific inhibitor of metalloproteinase ADAM8
  • IC50 value of 182 nM
  • Potential applications in inflammatory diseases
  • Potential applications in cancer
  • For research use only