Learn More
Medchemexpress LLC Cyclo(RLsKDK) TFA | 99.7% | 841.88 | 1 MG
SDP
Catalog No. 5000423667
Encompass_Preferred
Cyclo(RLsKDK) TFA is a specific inhibitor of metalloproteinase ADAM8 with an IC50 value of 182 nM. It is utilized in research for inflammatory diseases and cancer studies.
  • Specific inhibitor of metalloproteinase ADAM8 (IC50 182 nM).
  • Promotes ADAM8 activation and CD23 shedding (IC50 120 nM and 182 nM).
  • Increases activity of pro-ADAM8.
  • Reduces tumor load and improves survival rates in pancreatic ductal adenocarcinoma (PDAC) mice models.
  • Reduces sADAM8 content, pERK1/2 activation, and PDAC metastasis in vivo.
Catalog No. 50-004-23667 Supplier Medchemexpress LLC Supplier No. HYP1676A1MG
Cyclo(RLsKDK) TFA is a specific inhibitor of metalloproteinase ADAM8 with an IC50 value of 182 nM. It is utilized in research for inflammatory diseases and cancer studies.
  • Specific inhibitor of metalloproteinase ADAM8 (IC50 182 nM).
  • Promotes ADAM8 activation and CD23 shedding (IC50 120 nM and 182 nM).
  • Increases activity of pro-ADAM8.
  • Reduces tumor load and improves survival rates in pancreatic ductal adenocarcinoma (PDAC) mice models.
  • Reduces sADAM8 content, pERK1/2 activation, and PDAC metastasis in vivo.