Learn More
Cayman Chemical Cycloastragenol, Purity: ≥98%, CAS: 84605-18-5, 100 mg
SDP
Catalog No. 502951760
Encompass_Preferred
A telomerase activator that can slow telomere loss, increase replicative capacity, and improve immune function in CD8+ T lymphocytes from HIV-1-infected patients
Catalog No. 50-295-1760 Supplier Cayman Chemical Supplier No. 11722100 MG
A telomerase activator that can slow telomere loss, increase replicative capacity, and improve immune function in CD8+ T lymphocytes from HIV-1-infected patients