Learn More
AdipoGen Cycloastragenol
SDP
Supplier:  AdipoGen CDXC0592M050
Encompass_Preferred
Chemical. CAS 84605-18-5. Formula C30H50O5. MW 490.73. Cycloastragenol is an aglycone of astragaloside IV isolated from Astragalus species. Extensive pharmacological effects have been attributed to cycloastragenol, including telomerase activation, telomere elongation, anti-inflammatory and anti-oxidative properties, improvement of lipid metabolism and wound repair promotion. It has been shown to inhibit endoplasmic reticulum stress-associated TXNIP/NLRP3 inflammasome activation and NLRP3 inflammasome-mediated pyroptosis. It improved immune function in CD8 T lymphocytes from HIV-1-infected patients. It might be a novel candidate for age-associated diseases.
Catalog No. 50-256-1357