Learn More
Medchemexpress LLC Cyclobenzaprine (hydrochloride) | 6202-23-9 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0740R
Encompass_Preferred
Cyclobenzaprine hydrochloride is an orally active 5-HT2 receptor antagonist that centrally provides skeletal muscle relaxation, alleviates muscle spasms, and reduces pain. It also possesses antiparasitic activity, showing promise for research in acute, painful skeletal muscle disorders and infectious diseases.
  • Orally active 5-HT2 receptor antagonist
  • Provides skeletal muscle relaxation and alleviates muscle spasms
  • Reduces pain
  • Exhibits antiparasitic activity
  • Active against Leishmania infantum promastigotes and intracellular amastigotes
  • Induces oxidative stress in Leishmania infantum
  • Suppresses quadriceps activity in ischemic spinal cord rigidity models
  • Prevents tonic-extensor seizures
  • Reduces parasite load in Leishmania infantum-infected mice
  • Blocks α-motoneuronal excitation
Catalog No. 50-004-50763