Learn More
Medchemexpress LLC Cyclobenzaprine hydrochloride | 6202-23-9 | MFCD00079039 | 99.95% | 1 ML
SDP
Catalog No. 5000408152
Encompass_Preferred
Cyclobenzaprine hydrochloride is an orally active 5-HT2 receptor antagonist. It acts centrally to provide skeletal muscle relaxation, alleviate muscle spasms, and reduce pain. It also possesses antiparasitic activity and holds promise for research in acute, painful skeletal muscle disorders and infectious diseases.
  • Orally active 5-HT2 receptor antagonist
  • Provides skeletal muscle relaxation
  • Alleviates muscle spasms
  • Reduces pain
  • Possesses antiparasitic activity
  • Potential for research in acute, painful skeletal muscle disorders
  • Potential for research in infectious diseases
Catalog No. 50-004-08152 Supplier Medchemexpress LLC Supplier No. HYB074010MM/1ML
Cyclobenzaprine hydrochloride is an orally active 5-HT2 receptor antagonist. It acts centrally to provide skeletal muscle relaxation, alleviate muscle spasms, and reduce pain. It also possesses antiparasitic activity and holds promise for research in acute, painful skeletal muscle disorders and infectious diseases.
  • Orally active 5-HT2 receptor antagonist
  • Provides skeletal muscle relaxation
  • Alleviates muscle spasms
  • Reduces pain
  • Possesses antiparasitic activity
  • Potential for research in acute, painful skeletal muscle disorders
  • Potential for research in infectious diseases